2-Chloro-5-(difluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
PC49045-10G
PC49045-1G
PC49045-5G
APOSPC49045-10GEA
1100
EUR
APOSPC49045-10G
APOSPC49045-1G
APOSPC49045-5G
2-Chloro-5-(difluoromethyl)pyridine 97%
2-Chloro-5-(difluoromethyl)pyridine
Formel:
C₆H₄ClF₂N Molecular Weight: 163.56 g/mol Siedepunkt: 156…164 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD16140199 CAS-Nummer: 71701-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...