5-Chlor-2-(trifluormethyl)anilin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-chlorobenzotrifluoride

PC49064-5G PC49064-10G PC49064-1G
APOSPC49064-5GEA 89.9 EUR
APOSPC49064-5G APOSPC49064-10G APOSPC49064-1G
5-Chlor-2-(trifluormethyl)anilin 95%
5-Chlor-2-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
Molecular Weight: 195,5700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08282771
CAS-Nummer: 445-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR