4-Fluor-2-methoxyanilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC49081-100G
PC49081-25G
APOSPC49081-100GEA
105
EUR
APOSPC49081-100G
APOSPC49081-25G
4-Fluor-2-methoxyanilin 97%
4-Fluoro-2-methoxyaniline
Formel:
C₇H₈FNO Molecular Weight: 141,14 g/mol Schmelzpunkt: 45…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077536 CAS-Nummer: 450-91-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...