2-Chloro-3-[3-(trifluoromethyl)phenyl]quinoline 97%
Lieferant: Apollo Scientific
PC4911-1G
PC4911-500MG
PC4911-5G
APOSPC4911-1GEA
533
EUR
APOSPC4911-1G
APOSPC4911-500MG
APOSPC4911-5G
2-Chloro-3-[3-(trifluoromethyl)phenyl]quinoline 97%
2-Chloro-3-[3-(trifluoromethyl)phenyl]quinoline
Formel:
C₁₆H₉ClF₃N Schmelzpunkt: 102…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793215 CAS-Nummer: 339013-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...