1-Boc-3-fluoro-4-piperidone 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, tert-Butyl 3-Fluoro-4-oxo-1-piperidinecarboxylate, tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate, 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone, 3-Fluoro-4-oxo-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-fluoro-4-piperidone
PC49128-25G
PC49128-5G
PC49128-1G
APOSPC49128-25GEA
279
EUR
APOSPC49128-25G
APOSPC49128-5G
APOSPC49128-1G
1-Boc-3-fluoro-4-piperidone 96%
1-Boc-3-fluoro-4-piperidone
Formel:
C₁₀H₁₆FNO₃ Molecular Weight: 217,24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD10566554 CAS-Nummer: 211108-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...