4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoic acid 97%
Lieferant: Apollo Scientific
PC4913-1G
PC4913-500MG
PC4913-5G
APOSPC4913-1GEA
275
EUR
APOSPC4913-1G
APOSPC4913-500MG
APOSPC4913-5G
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoic acid 97%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoic acid
Formel:
C₁₀H₁₀ClF₃N₂O₂ Molecular Weight: 282,65 g/mol Schmelzpunkt: 92…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172556 CAS-Nummer: 338770-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...