1,4-Dibrom-3,5-difluorbenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dibromo-1,3-difluorobenzene

PC49154-1G PC49154-5G PC49154-25G
APOSPC49154-1GEA 21.9 EUR
APOSPC49154-1G APOSPC49154-5G APOSPC49154-25G
1,4-Dibrom-3,5-difluorbenzol
1,4-Dibromo-3,5-difluorobenzene
Formel: C₆H₂Br₂F₂
Molecular Weight: 271,8849 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07783665
CAS-Nummer: 128259-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR