Methyl-6-fluornicotinat 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate
PC49205-25G
PC49205-5G
APOSPC49205-25GEA
227
EUR
APOSPC49205-25G
APOSPC49205-5G
Methyl-6-fluornicotinat 98%
Methyl-6-fluornicotinat
Formel:
C₇H₆FNO₂ Molecular Weight: 155.13 g/mol Schmelzpunkt: 48…52 °C Flammpunkt: 98.9 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095098 CAS-Nummer: 1427-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...