4-Chlor-3-(trifluormethoxy)anilin

Lieferant: Apollo Scientific
Achtung

PC49252-1G PC49252-5G PC49252-250MG
APOSPC49252-1GEA 0 EUR
APOSPC49252-1G APOSPC49252-5G APOSPC49252-250MG
4-Chlor-3-(trifluormethoxy)anilin
4-Chlor-3-(trifluormethoxy)anilin
Formel: C₇H₅ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09832255
CAS-Nummer: 97608-50-9
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR