3-Amino-2-fluorbenzylalkohol 97%

Lieferant: Apollo Scientific

Synonyme: (3-Amino-2-fluorophenyl)methanol

PC49265-1G PC49265-250MG
APOSPC49265-1GEA 228 EUR
APOSPC49265-1G APOSPC49265-250MG
3-Amino-2-fluorbenzylalkohol 97%
3-Amino-2-fluorbenzylalkohol
Formel: C₇H₈FNO
Molecular Weight: 141,1429 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11846488
CAS-Nummer: 1051899-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR