3-Methoxy-5-(trifluormethyl)benzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

PC49338-10G PC49338-5G PC49338-1G
APOSPC49338-10GEA 406 EUR
APOSPC49338-10G APOSPC49338-5G APOSPC49338-1G
3-Methoxy-5-(trifluormethyl)benzylalkohol 97%
3-Methoxy-5-(trifluormethyl)benzylalkohol
Formel: C₉H₉F₃O₂
Molecular Weight: 206.16 g/mol
Schmelzpunkt: 32…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025413
CAS-Nummer: 916420-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR