3-Methoxy-5-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC49339-10G PC49339-5G PC49339-1G
APOSPC49339-10GEA 0 EUR
APOSPC49339-10G APOSPC49339-5G APOSPC49339-1G
3-Methoxy-5-(trifluormethyl)benzylbromid 98%
3-Methoxy-5-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09025414
CAS-Nummer: 916421-00-6
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR