Methyl-2-fluoro-4-iodonicotinate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-fluoro-4-iodonicotinate
PC49357-250MG
PC49357-5G
APOSPC49357-250MGEA
21.9
EUR
APOSPC49357-250MG
APOSPC49357-5G
Methyl-2-fluoro-4-iodonicotinate 97%
Methyl-2-fluoro-4-iodonicotinate
Formel:
C₇H₅FINO₂ Molecular Weight: 281,0200 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04972401 CAS-Nummer: 884494-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...