3-Fluoro-4-iodobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC49375-1G PC49375-5G PC49375-10G
APOSPC49375-1GEA 34.8 EUR
APOSPC49375-1G APOSPC49375-5G APOSPC49375-10G
3-Fluoro-4-iodobenzaldehyde
3-Fluoro-4-iodobenzaldehyde
Formel: C7H4FIO
Molecular Weight: 250,0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07782055
CAS-Nummer: 1003709-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR