4-Fluor-m-anisaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluor-3-anisaldehyd, 4-Fluor-3-methoxybenzaldehyd
PC4939-25G
PC4939-5G
PC4939-100G
APOSPC4939-25GEA
89.9
EUR
APOSPC4939-25G
APOSPC4939-5G
APOSPC4939-100G
4-Fluor-m-anisaldehyd 98%
4-Fluor-m-anisaldehyd
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol Schmelzpunkt: 58…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143320 CAS-Nummer: 128495-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...