Ethyl-6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate

PC49398-1G PC49398-250MG
APOSPC49398-1GEA 126 EUR
APOSPC49398-1G APOSPC49398-250MG
Ethyl-6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₀H₈BrFN₂O₂
Molecular Weight: 287,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13195312
CAS-Nummer: 1260763-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR