Ethyl-4,4,4-trifluoracetoacetat 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4,4-Trifluoracetessigsäureethylester, ω,ω,ω-Trifluoracetessigsäureethylester, 4,4,4-Trifluor-3-oxobuttersäureethylester
PC49444-100G
PC49444-1KG
PC49444-500G
APOSPC49444-100GEA
37.7
EUR
APOSPC49444-100G
APOSPC49444-1KG
APOSPC49444-500G
Ethyl-4,4,4-trifluoracetoacetat 90%
Ethyl-4,4,4-trifluoracetoacetat
Formel:
C₆H₇F₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000424 CAS-Nummer: 372-31-6 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...