1,1,1,2-Tetrafluor-4-iod-2-(trifluormethyl)butan 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4,4,4-Tetrafluoro-3-(trifluoromethyl)butyl Iodide

PC4945-1G PC4945-5G
APOSPC4945-1GEA 29 EUR
APOSPC4945-1G APOSPC4945-5G
1,1,1,2-Tetrafluor-4-iod-2-(trifluormethyl)butan 97%
1,1,1,2-Tetrafluoro-4-iodo-2-(trifluoromethyl)butane
Formel: C₅H₄F₇I
Molecular Weight: 323,98 g/mol
Siedepunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042316
CAS-Nummer: 99324-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR