3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
Lieferant: Apollo Scientific
|
Achtung
|
PC4948-1G
PC4948-5G
APOSPC4948-1GEA
92.8
EUR
APOSPC4948-1G
APOSPC4948-5G
3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
Formel:
C₄H₂ClF₆I Molecular Weight: 326,41 g/mol Siedepunkt: 124…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093304 CAS-Nummer: 240122-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...