5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carboxylic acid

Lieferant: Apollo Scientific

PC4951-1G PC4951-500MG
APOSPC4951-1GEA 1060 EUR
APOSPC4951-1G APOSPC4951-500MG
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carboxylic acid
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carboxylic acid
Formel: C₁₁H₉ClF₃N₃O₃
Molecular Weight: 323,66 g/mol
Schmelzpunkt: 161…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202620
CAS-Nummer: 251097-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR