2,3,6-Trifluorobenzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC49545-1G
PC49545-5G
PC49545-10G
APOSPC49545-1GEA
88.6
EUR
APOSPC49545-1G
APOSPC49545-5G
APOSPC49545-10G
2,3,6-Trifluorobenzenesulphonyl chloride 97%
2,3,6-Trifluorobenzenesulphonyl chloride
Formel:
C₆H₂ClF₃O₂S Siedepunkt: 66…68 °C/0,1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832391 CAS-Nummer: 1017779-75-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...