Lanthan(III)trifluormethansulfonat 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate

PC4963-5G PC4963-25G PC4963-100G
APOSPC4963-5GEA 68.3 EUR
APOSPC4963-5G APOSPC4963-25G APOSPC4963-100G
Lanthan(III)trifluormethansulfonat 98%
Lanthan(III)trifluormethansulfonat
Formel: C₃F₉LaO₉S₃
Molecular Weight: 586,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077560
CAS-Nummer: 52093-26-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR