2-Fluor-4-(trifluormethoxy)anilin 99%

Lieferant: Apollo Scientific
Gefahr

PC49655-1G PC49655-5G
APOSPC49655-1GEA 174 EUR
APOSPC49655-1G APOSPC49655-5G
2-Fluor-4-(trifluormethoxy)anilin 99%
2-Fluor-4-(trifluormethoxy)anilin
Formel: C₇H₅F₄NO
Molecular Weight: 195,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527448
CAS-Nummer: 123572-58-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR