2,4,5-Trifluorbenzonitril 96%

Lieferant: Apollo Scientific

PC49703-100G PC49703-25G PC49703-5G
APOSPC49703-100GEA 156 EUR
APOSPC49703-100G APOSPC49703-25G APOSPC49703-5G
2,4,5-Trifluorbenzonitril 96%
2,4,5-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013289
CAS-Nummer: 98349-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR