4-Chlor-2-fluorphenylacetonitril 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-2-fluorobenzeneacetonitrile
PC49721-100G
PC49721-25G
PC49721-5G
APOSPC49721-100GEA
191
EUR
APOSPC49721-100G
APOSPC49721-25G
APOSPC49721-5G
4-Chlor-2-fluorphenylacetonitril 95%
4-Chlor-2-fluorphenylacetonitril
Formel:
C₈H₅ClFN Molecular Weight: 169,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01312758 CAS-Nummer: 75279-53-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...