4-Fluor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-3-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 4-Fluoro-3-(pyrrolidine-1-carbonyl)phenylboronic acid
PC4989-1G
PC4989-5G
APOSPC4989-1GEA
229
EUR
APOSPC4989-1G
APOSPC4989-5G
4-Fluor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%
4-Fluor-3-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₃BFNO₃ Schmelzpunkt: 152…154 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235050 CAS-Nummer: 874219-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...