(2-Methyl-3-(trifluormethyl)phenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

PC500183-1G PC500183-5G
APOSPC500183-1GEA 145 EUR
APOSPC500183-1G APOSPC500183-5G
(2-Methyl-3-(trifluormethyl)phenyl)essigsäure
(2-Methyl-3-(trifluormethyl)phenyl)essigsäure
Formel: C₁₀H₉F₃O₂
Molecular Weight: 218,18 g/mol
Schmelzpunkt: 105…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832279
CAS-Nummer: 1000546-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR