2-(5-Bromo-2-fluorophenyl)acetonitrile 98%

Lieferant: Apollo Scientific
Achtung

PC500188-10G PC500188-5G
APOSPC500188-10GEA 101 EUR
APOSPC500188-10G APOSPC500188-5G
2-(5-Bromo-2-fluorophenyl)acetonitrile 98%
2-(5-Bromo-2-fluorophenyl)acetonitrile
Formel: C₈H₅BrFN
Molecular Weight: 214,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09804785
CAS-Nummer: 305800-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR