3-Brom-6-fluorimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC50020-1G
PC50020-250MG
PC50020-5G
APOSPC50020-1GEA
106
EUR
APOSPC50020-1G
APOSPC50020-250MG
APOSPC50020-5G
3-Brom-6-fluorimidazo[1,2-a]pyridin
3-Brom-6-fluorimidazo[1,2-a]pyridin
Formel:
C₇H₄BrFN₂ Molecular Weight: 215,200 g/mol Schmelzpunkt: 141…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827770 CAS-Nummer: 1186405-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...