2-Brom-4-fluor-5-nitrophenol 97%

Lieferant: Apollo Scientific
Achtung

PC5004-25G PC5004-5G PC5004-100G
APOSPC5004-25GEA 81.2 EUR
APOSPC5004-25G APOSPC5004-5G APOSPC5004-100G
2-Brom-4-fluor-5-nitrophenol 97%
2-Brom-4-fluor-5-nitrophenol
Formel: C₆H₃BrFNO₃
Molecular Weight: 236,00 g/mol
Schmelzpunkt: 126…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08460104
CAS-Nummer: 84478-87-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR