3'-Brom-2,2,2,2'-tetrafluoracetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanone

PC50065-1G PC50065-250MG
APOSPC50065-1GEA 580 EUR
APOSPC50065-1G APOSPC50065-250MG
3'-Brom-2,2,2,2'-tetrafluoracetophenon 97%
3'-Brom-2,2,2,2'-tetrafluoracetophenon
Formel: C₈H₃BrF₄O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 871353-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR