2,3,4-Trifluorbenzylamin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2,3,4-Trifluorophenyl)methanamine

PC5008-10G PC5008-5G PC5008-1G
APOSPC5008-10GEA 261 EUR
APOSPC5008-10G APOSPC5008-5G APOSPC5008-1G
2,3,4-Trifluorbenzylamin 98%
2,3,4-Trifluorbenzylamin
Formel: C₇H₆F₃N
Molecular Weight: 161.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236313
CAS-Nummer: 235088-67-2
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR