2-Amino-4-bromo-5-(trifluoromethyl)phenylamine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-5-(trifluoromethyl)benzene-1,2-diamine

PC501059-1G PC501059-250MG PC501059-5G
APOSPC501059-1GEA 377 EUR
APOSPC501059-1G APOSPC501059-250MG APOSPC501059-5G
2-Amino-4-bromo-5-(trifluoromethyl)phenylamine 95%
2-Amino-4-bromo-5-(trifluoromethyl)phenylamine
Formel: C₇H₆BrF₃N₂
Molecular Weight: 255.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263998
CAS-Nummer: 157590-60-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR