2'-Fluor-3'-nitroacetophenon 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-3-nitrophenyl)ethanone

PC501170-1G PC501170-5G PC501170-10G PC501170-25G
APOSPC501170-1GEA 89.9 EUR
APOSPC501170-1G APOSPC501170-5G APOSPC501170-10G APOSPC501170-25G
2'-Fluor-3'-nitroacetophenon 95%
2'-Fluor-3'-nitroacetophenon
Formel: C₈H₆FNO₃
Molecular Weight: 183,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194361
CAS-Nummer: 873697-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR