1-Brom-2-fluor-4-(methylsulfonyl)benzol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Bromo-3-fluorophenyl methyl sulphone

PC501361-10G PC501361-1G PC501361-5G PC501361-25G
APOSPC501361-10GEA 575 EUR
APOSPC501361-10G APOSPC501361-1G APOSPC501361-5G APOSPC501361-25G
1-Brom-2-fluor-4-(methylsulfonyl)benzol
1-Bromo-2-fluoro-4-(methylsulphonyl)benzene
Formel: C₇H₆BrFO₂S
Molecular Weight: 253.0887 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053812
CAS-Nummer: 1032825-02-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR