3-Fluor-5-nitrobenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC501430-5G
APOSPC501430-5GEA
172
EUR
APOSPC501430-5G
3-Fluor-5-nitrobenzaldehyd 97%
3-Fluoro-5-nitrobenzaldehyde
Formel:
C₇H₄FNO₃ Molecular Weight: 169,1100 g/mol Schmelzpunkt: 47…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111000 CAS-Nummer: 108159-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...