4-Brom-2-chlor-5-fluor-1-iodbenzol 95%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2-chloro-5-fluoroiodobenzene, 1-Bromo-5-chloro-2-fluoro-4-iodobenzene
PC501466-1G
PC501466-5G
PC501466-25G
APOSPC501466-1GEA
87
EUR
APOSPC501466-1G
APOSPC501466-5G
APOSPC501466-25G
4-Brom-2-chlor-5-fluor-1-iodbenzol 95%
4-Brom-2-chlor-5-fluor-1-iodbenzol
Formel:
C₆H₂BrClFI Molecular Weight: 335,34 g/mol Schmelzpunkt: 34…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878157 CAS-Nummer: 1000572-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...