2-Chloro-7-fluoro-1H-benzo[d]imidazole
Lieferant: Apollo Scientific
|
Achtung
|
PC501474-1G
PC501474-250MG
PC501474-5G
APOSPC501474-1GEA
203
EUR
APOSPC501474-1G
APOSPC501474-250MG
APOSPC501474-5G
2-Chloro-7-fluoro-1H-benzo[d]imidazole
2-Chloro-7-fluoro-1H-benzo[d]imidazole
Formel:
C₇H₄ClFN₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
256519-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...