Methyl-2-fluoro-5-formylbenzoate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl-2-Fluoro-5-formyl-benzoate

PC501609-1G PC501609-5G PC501609-10G
APOSPC501609-1GEA 42.2 EUR
APOSPC501609-1G APOSPC501609-5G APOSPC501609-10G
Methyl-2-fluoro-5-formylbenzoate 95%
Methyl-2-fluoro-5-formylbenzoate
Formel: C₉H₇FO₃
Molecular Weight: 182.15 g/mol
Schmelzpunkt: 62…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18904319
CAS-Nummer: 165803-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR