Methyl-2-fluoro-4-formylbenzoate 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-fluoro-4-formylbenzoate

PC501652-1G PC501652-5G
APOSPC501652-1GEA 33.5 EUR
APOSPC501652-1G APOSPC501652-5G
Methyl-2-fluoro-4-formylbenzoate 96%
Methyl-2-fluoro-4-formylbenzoate
Formel: C₉H₇FO₃
Molecular Weight: 182.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20231454
CAS-Nummer: 85070-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR