5-Brom-2,4-difluortoluol

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-2,4-difluoro-5-methylbenzene

PC501997-1G PC501997-25G PC501997-5G
APOSPC501997-1GEA 21.9 EUR
APOSPC501997-1G APOSPC501997-25G APOSPC501997-5G
5-Brom-2,4-difluortoluol
5-Brom-2,4-difluortoluol
Formel: C₇H₅BrF₂
Molecular Weight: 207,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15144620
CAS-Nummer: 159277-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR