5-Fluoro-2-iodobenzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC50205-1G PC50205-250MG PC50205-5G
APOSPC50205-1GEA 112 EUR
APOSPC50205-1G APOSPC50205-250MG APOSPC50205-5G
5-Fluoro-2-iodobenzaldehyde 97%
5-Fluoro-2-iodobenzaldehyde
Formel: C₇H₄FIO
Lagertemperatur: Raumtemperatur
MDL: MFCD07782057
CAS-Nummer: 877264-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR