1,2,5-Trifluor-3-nitrobenzol

Lieferant: Apollo Scientific

PC502429-1G PC502429-250MG PC502429-5G
APOSPC502429-1GEA 256 EUR
APOSPC502429-1G APOSPC502429-250MG APOSPC502429-5G
1,2,5-Trifluor-3-nitrobenzol
1,2,5-Trifluor-3-nitrobenzol
Formel: C₆H₂F₃NO₂
Molecular Weight: 177,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00456804
CAS-Nummer: 66684-57-9
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR