4-Brom-5-chlor-2-fluoranilin

Lieferant: Apollo Scientific

PC502456-10G PC502456-1G PC502456-5G
APOSPC502456-10GEA 655 EUR
APOSPC502456-10G APOSPC502456-1G APOSPC502456-5G
4-Brom-5-chlor-2-fluoranilin
4-Brom-5-chlor-2-fluoranilin
Formel: C₆H₄BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD09878173
CAS-Nummer: 116369-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR