Methyl-3-fluor-2-methoxybenzoat

Lieferant: Apollo Scientific

Synonyme: 3-Fluor-2-methoxybenzoesäuremethylester

PC502591-1G PC502591-5G
APOSPC502591-1GEA 109 EUR
APOSPC502591-1G APOSPC502591-5G
Methyl-3-fluor-2-methoxybenzoat
Methyl-3-fluor-2-methoxybenzoat
Formel: C₉H₉FO₃
Molecular Weight: 184,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09753133
CAS-Nummer: 106428-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR