Methyl-2-fluoro-6-nitrobenzoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-fluoro-6-nitrobenzoate
PC502597-1G
APOSPC502597-1GEA
81.2
EUR
APOSPC502597-1G
Methyl-2-fluoro-6-nitrobenzoate
Methyl-2-fluoro-6-nitrobenzoate
Formel:
C₈H₆FNO₄ Molecular Weight: 199,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09880026 CAS-Nummer: 212189-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...