4-Fluor-3-(2-hydroxyethylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-3-(2-hydroxyethylcarbamoyl)benzeneboronic acid
PC5028-1G
PC5028-5G
APOSPC5028-1GEA
273
EUR
APOSPC5028-1G
APOSPC5028-5G
4-Fluor-3-(2-hydroxyethylcarbamoyl)phenylboronsäure 98%
4-Fluor-3-(2-hydroxyethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₁BFNO₄ Molecular Weight: 227,00 g/mol Schmelzpunkt: 89…91 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235044 CAS-Nummer: 874219-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...