4-Brom-2-fluorphenylisothiocyanat
Lieferant: Apollo Scientific
|
Gefahr
|
PC503014-25G
PC503014-5G
APOSPC503014-25GEA
261
EUR
APOSPC503014-25G
APOSPC503014-5G
4-Brom-2-fluorphenylisothiocyanat
4-Brom-2-fluorphenylisothiocyanat
Formel:
C₇H₃BrFNS Molecular Weight: 232,08 g/mol Schmelzpunkt: 37…42 °C Flammpunkt: 110 °C Lagertemperatur: Kühlschrank |
CAS-Nummer:
81171-71-3 UN: 1760 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...