4-Brom-2-fluorphenylisocyanat

Lieferant: Apollo Scientific
Gefahr

PC503015-1G PC503015-25G PC503015-5G
APOSPC503015-1GEA 56.7 EUR
APOSPC503015-1G APOSPC503015-25G APOSPC503015-5G
4-Brom-2-fluorphenylisocyanat
4-Brom-2-fluorphenylisocyanat

  • May form small amount of sediment.

Formel: C₇H₃BrFNO
Molecular Weight: 216,01 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 88112-75-8
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR