3-Cyan-4-fluoranilin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril
PC50393-100G
PC50393-25G
APOSPC50393-100GEA
149
EUR
APOSPC50393-100G
APOSPC50393-25G
3-Cyan-4-fluoranilin 95%
3-Cyan-4-fluoranilin
Formel:
C₇H₅FN₂ Molecular Weight: 136.13 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00277872 CAS-Nummer: 53312-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...